Skip to Content
MilliporeSigma

MTCT8309

MTC™ Bio Culture Tube Cap

two position, polyethylene, for use with 12 x 75 mm tubes, pack of 1000 ea (8 x 250ea)

Synonym(s):

push cap, tube closure, MTC Bio T8309, cap, closure

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41121800
Material:
cap (snap), clear , polyethylene
Sterility:
sterile
Packaging:
pack of 1000 ea (8 x 250ea)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

cap (snap), clear , polyethylene

sterility

sterile

packaging

pack of 1000 ea (8 x 250ea)

manufacturer/tradename

MTC™ Bio T8309

General description

Cap, two-position, for 12×75mm tubes, PE, 125/bag, 1000/cs

Application

cell culture

Features and Benefits

  • for 12 mm x 75 mm tubes
  • two position locking
  • sterilized to SAL 10-6
  • RNase, DNase, pyrogen and cytotoxin free

Legal Information

MTC is a trademark of MTC Bio


Still not finding the right product?

Explore all of our products under MTC™ Bio Culture Tube Cap




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library