Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C19H20BrN2O4P
CAS Number:
Molecular Weight:
451.25
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
eCl@ss:
32160702
Assay:
≥98% (HPLC)
Form:
powder
assay
≥98% (HPLC)
Quality Segment
form
powder
color
off-white to pale yellow
solubility
DMSO: ≥20 mg/mL
storage temp.
2-8°C
SMILES string
CCOP(=O)(Cc1ccc(cc1)C(=O)Nc2ccc(Br)cc2C#N)OCC
InChI
1S/C19H20BrN2O4P/c1-3-25-27(24,26-4-2)13-14-5-7-15(8-6-14)19(23)22-18-10-9-17(20)11-16(18)12-21/h5-11H,3-4,13H2,1-2H3,(H,22,23)
InChI key
KPRTURMJVWXURQ-UHFFFAOYSA-N
Biochem/physiol Actions
Lipoprotein lipase activator. Overexpression of lipoprotein lipase in transgenic rabbits leads to increased small dense LDL in plasma and promotes atherosclerosis. Long-term administration of NO-1886 protects against the development of experimental atherosclerosis in animals.
Packaging
Packaged under nitrogen
Signal Word
Warning
Hazard Codes
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Storage Class
11 - Combustible Solids
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
