Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.41
UNSPSC Code:
12352203
biological source
goat
Quality Segment
antibody form
affinity purified immunoglobulin
antibody product type
primary antibodies
species reactivity
human
technique(s)
immunoprecipitation (IP): 5- 20 μg/mg, western blot: 1:1,000- 1:10,000
accession no.
NP_001265.1
shipped in
wet ice
storage temp.
2-8°C
Gene Information
goat ... Chk1(1111)
Immunogen
The epitope recognized by PLA0055 maps to a region between residues 250 and 300 of human Checkpoint Kinase 1 using the numbering given in entry NP_001265.1 (GeneID 1111).
Physical form
Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% sodium azide
Other Notes
Chk1 functions as a checkpoint protein kinase that is critical to the regulation of the Cdc25a and Cdc25c phosphatases that promote S and G2/M phase progression, respectively. Chk1 kinase activity inhibits cell cycle progression in the presence of DNA damage by phosphorylating Cdc25a and Cdc25c and causing the degradation of Cdc25a and cytoplasmic sequestration of Cdc25c.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
WGK
nwg
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.