Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.32
UNSPSC Code:
12352200
species reactivity
mouse, rat, human
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable, enzyme immunoassay: suitable
input
sample type culture supernatant(s)
sample type plasma
sample type cell lysate
sample type serum
assay range
inter-assay cv: <15%
intra-assay cv: <10%
sensitivity: 161 pg/mL
standard curve range: 0.1-1000 ng/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... GHRL(51738)
General description
The Ghrelin Enzyme Immunoassay (EIA) Kit is an in vitro quantitative assay for detecting Ghrelin peptide based on the principle of Competitive Enzyme Immunoassay.
Immunogen
Ghrelin, synthetic peptide
Application
For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Signal Word
Warning
Hazard Codes
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class
10 - Combustible liquids
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
