Skip to Content
MilliporeSigma

SAB4501554

Anti-INSL4 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

EPIL, Early placenta insulin-like peptide, Insulin-like peptide 4, Placentin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ELISA, IF, WB
Citations:
2
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 15 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000, immunofluorescence: 1:100-1:500, western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... INSL4(3641)

General description

Early placental insulin-like protein (INSL4 or EPIL) belongs to the insulin superfamily of hormones. It is expressed at high level in the placenta. The gene encoding INSL4 is located on human chromosome 9p24 and consists of two exons and one intron.
Anti-INSL4 antibody detects endogenous levels of total INSL4 protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human INSL4.

Immunogen Range: 41-90

Biochem/physiol Actions

Early placental insulin-like protein (INSL4 or EPIL) helps in the development of placental and fetal membrane. It may play a major role in the progression trophoblast and regulation of bone formation.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

— or —

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

WGK

nwg

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library