Skip to Content
MilliporeSigma

SAB4702011

Monoclonal Anti-TCR gamma/delta purified low endotoxin produced in mouse

Synonym(s):

TCRG/D

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Conjugate:
unconjugated
Clone:
B1, monoclonal
Application:
FACS, FUNC, IHC
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


conjugate

unconjugated

Quality Segment

antibody product type

primary antibodies

clone

B1, monoclonal

form

buffered aqueous solution

species reactivity

human, ( Non-human primates)

concentration

1 mg/ml

technique(s)

FUNC: suitable, flow cytometry: suitable, immunohistochemistry: suitable

isotype

IgG1κ

storage temp.

2-8°C

target post-translational modification

unmodified

General description

The antigen-specific T cell receptor (TCR) is composed of either alpha and beta subunit, or gamma and delta subunit. Majority of T cells present in the blood, lymph and secondary lymphoid organs express TCR alpha/beta heterodimers, whereas the T cells expressing TCR gamma/delta heterodimers are localized mainly in epithelial tissues and at the sites of infection. The subunits of TCR heterodimers are covalently bonded and in the endoplasmic reticulum they associate with CD3 subunits to form functional TCR-CD3 complex. Lack of expression of any of the chains is sufficient to stop cell surface expression.

Biochem/physiol Actions

The mouse monoclonal antibody B1 (also known as B1.1) recognizes an extracellular epitope of TCR gamma/delta, the subtype of T cell receptor expressed mainly in epithelial tissues and at the sites of infection.

Physical form

Phosphate buffered saline (PBS), pH 7.4

Preparation Note

Purified by protein-A affinity chromatography.
Store at 2-8°C. Do not freeze.

Other Notes

The antibody solution should be gently mixed before use.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

WGK

WGK 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library