Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
Conjugate:
Peridinin-Chlorophyll-Protein Complex conjugate
Clone:
BU63, monoclonal
Application:
FACS
Citations:
-
conjugate
Peridinin-Chlorophyll-Protein Complex conjugate
Quality Segment
antibody product type
primary antibodies
clone
BU63, monoclonal
form
buffered aqueous solution
species reactivity
human
technique(s)
flow cytometry: suitable
isotype
IgG1
UniProt accession no.
storage temp.
2-8°C
target post-translational modification
unmodified
Gene Information
human ... CD86(942)
General description
CD80 (B7-1) and CD86 (B7-2) are ligands of T cell critical costimulatory molecule CD28 and of an inhibitory receptor CTLA-4 (CD152). The both B7 molecules are expressed on professional antigen-presenting cells and are essential for T cell activation, the both molecules can also substitute for each other in this process. The question what are the differences in CD80 and CD86 competency has not been fully elucidated yet; there are still conflicts in results about their respective roles in initiation or sustaining of the T cell immune response.
Immunogen
B-lymphoblastoid cell line ARH 77
Biochem/physiol Actions
The mouse monoclonal antibody BU63 reacts with an extracellular epitope of CD86 (B7-2), a 70 kDa type I transmembrane glycoprotein of immunoglobulin supergene family, expressed on professional antigen-presenting cells, such as dendritic cells, macrophages or activated B lymphocytes.
Physical form
Stabilizing phosphate buffered saline (PBS), pH 7.4, 15 mM sodium azide
Preparation Note
Purified antibody is conjugated with activated Peridinin-Chlorophyll Protein (PerCP) under optimum conditions and unconjugated antibody and free fluorochrome are removed by size-exclusion chromatography.
Store at 2-8°C. Protect from prolonged exposure to light. Do not freeze.
Other Notes
The antibody solution should be gently mixed before use.
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.