Skip to Content
MilliporeSigma

SAB5801037

Goat Anti-Mouse kappa Alpaca Heavy chain only Antibody

recombinant Alpaca(VHH) Nano antibody

Synonym(s):

Recombinant Alpaca single variable domain secondary Nanobody

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
recombinant monoclonal
Application:
ELISA, WB
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

alpaca

Quality Segment

recombinant

expressed in HEK 293 cells (Human cell line)

conjugate

unconjugated

antibody form

affinity purified immunoglobulin

antibody product type

secondary antibodies

clone

recombinant monoclonal

form

lyophilized solid

mol wt

100 kDa

species reactivity

mouse, Alpaca

technique(s)

ELISA: 1:5000-1:20000, western blot: 1:5000-1:20000

size

0.1 mL

color

colorless

isotype

VHH

shipped in

wet ice

storage temp.

2-8°C

General description

Goat Anti-Mouse kappa Alpaca Heavy chain only Antibody is designed to specifically detect mouse IgG kappa chain. It is a monovalent, single domain, recombinant antibody fused to goat IgG Fc. Based on immunoelectrophoresis and/or ELISA, Goat Anti-Mouse kappa Alpaca Heavy chain only Antibody reacts with the mouse IgG kappa chain selectively but not with the mouse IgG lambda chain or heavy chain of mouse IgG.

Most monoclonal antibodies are generated in mouse. There are five antibody isotypes (IgA, IgD, IgE, IgG, and IgM) from mouse. Each isotype has a different heavy chain. Mouse IgG constitutes 75% of serum immunoglobulins, and IgG is the predominant form of first antibody produced from mouse. Mouse IgG consists of five subclasses-IgG1, IgG2a, IgG2b, IgG2c(inbred mouse strains with the Igh1-b allele have IgG2c isotype instead of IgG2a), IgG3. They are highly homologous and differ mainly in the hinge region. The whole IgG molecule possesses both the Fc region and the Fab region, which possessing the epitope-recognition site. The IgG contains two heavy and light chains(kappa or lambda), and the heavy chain is about 50 KD and the light chain is about 25 KD. The common IgG is monomeric with a molecular weight of approximately 150 kD.

VHH are single-domain antibodies derived from the variable regions of heavy chain of Camelidae immunoglobulin. The size of VHH is extremely small(less than15KDa) compared to other forms of antibody fragment, which significantly increase the permeability of VHH. Thus VHH is considered of great value for research, diagnostics and therapeutics.

Immunogen

Recombinant Alpaca single variable domain secondary nano antibody

Biochem/physiol Actions

Mouse IgG kappa chain
Isotype: VHH domain of alpaca IgG2b/2c fused to goat IgG Fc

Packaging

100 μg in Glass bottle

Physical form

Lyophilized from 10mM PBS (pH 7.5), 0.05% sucrose, 0.1% trehalose

Preparation Note

Add 100uL dH2O or 100uL dH2O with 50% Glycerol to a final concentration of 1mg/mL
After reconstitution, store at –20 °C(Avoid freeze / thaw cycles). Stable for 12 months at -20°C

Other Notes

No cross-reactivity with rabbit, human, cynomolgus, rat, goat IgG


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

13 - Non Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library