Skip to Content
MilliporeSigma

SAB5801042

Goat Anti-Mouse IgM(µ chain) HRP Alpaca Heavy chain only Antibody

recombinant Alpaca(VHH) Nano antibody

Synonym(s):

Recombinant Alpaca single variable domain secondary Nanobody

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
horse radish peroxidase (HRP)
Clone:
recombinant monoclonal
Application:
ELISA, WB
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

alpaca

Quality Segment

recombinant

expressed in HEK 293 cells (Human cell line)

conjugate

horse radish peroxidase (HRP)

antibody form

affinity purified immunoglobulin

antibody product type

secondary antibodies

clone

recombinant monoclonal

form

lyophilized solid

mol wt

100 kDa

species reactivity

mouse, Alpaca

technique(s)

ELISA: 1:5000-1:20000, western blot: 1:5000-1:20000

size

0.1 mL

color

colorless

isotype

VHH

shipped in

wet ice

storage temp.

2-8°C

General description

Goat Anti-Mouse IgM(µ chain) HRP Alpaca Heavy chain only Antibody is designed to specifically detect mouse IgM. It is a monoclonal, recombinant, goat IgG Fc fused single domain antibody to µ chain of mouse IgM coupled to HRP. Based on immunoelectrophoresis and/or ELISA, Goat Anti-Mouse IgM(µ chain) HRP Alpaca Heavy chain only Antibody reacts with the µ chain of mouse IgM selectively but not with mouse, rabbit, human, cynomolgus, rat, goat IgG.

Most monoclonal antibodies are generated in mouse. There are five antibody isotypes (IgA, IgD, IgE, IgG, and IgM) from mouse. Each isotype has a different heavy chain. IgM accounts for 5-10% of the immunoglobulin pool and is the predominant antibody in the primary immune response. Unlike IgG, IgM does not contain a hinge region but does contain an additional constant domain. The monomeric form IgM has a molecular weight of 180 KD. It is classically represented as a pentamer of the basic four chain structure held together by a J chain but can also exist in a hexameric form without the J chain and as a monomer on the surface of B-cells.

VHH are single-domain antibodies derived from the variable regions of heavy chain of Camelidae immunoglobulin. The size of VHH is extremely small(less than15KDa) compared to other forms of antibody fragment, which significantly increase the permeability of VHH. Thus VHH is considered of great value for research, diagnostics and therapeutics.

Immunogen

Recombinant Alpaca single variable domain secondary nano antibody

Biochem/physiol Actions

Fc region of mouse IgM (µ chain specific)
Isotype: VHH domain of alpaca IgG2b/2c fused to goat IgG Fc

Packaging

100 μg in Glass bottle

Physical form

Lyophilized from 10mM PBS (pH 7.5), 0.05% sucrose, 0.1% trehalose

Preparation Note

Add 100uL dH2O or 100uL dH2O with 50% Glycerol to a final concentration of 1mg/mL
After reconstitution, store at –20 °C(Avoid freeze / thaw cycles). Protect from light. Stable for 12 months at -20°C

Other Notes

No cross-reactivity with mouse, rabbit, human, cynomolgus, rat, goat IgG


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

13 - Non Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library