Skip to Content
MilliporeSigma

SAB5801062

Rabbit Anti-Alpaca IgG2c Biotin Alpaca Heavy chain only Antibody

recombinant Alpaca(VHH) Nano antibody

Synonym(s):

Recombinant Alpaca single variable domain secondary Nanobody

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
biotin
Clone:
recombinant monoclonal
Application:
ELISA, WB
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

alpaca

Quality Segment

recombinant

expressed in HEK 293 cells (Human cell line)

conjugate

biotin

antibody form

affinity purified immunoglobulin

antibody product type

secondary antibodies

clone

recombinant monoclonal

form

lyophilized solid

mol wt

100 kDa

species reactivity

Alpaca, (alpaca)

technique(s)

ELISA: 1:4000-1:10000, western blot: 1:4000-1:10000

size

0.1 mL

color

colorless

isotype

VHH

shipped in

wet ice

storage temp.

2-8°C

General description

Rabbit Anti-Alpaca IgG2c Biotin Alpaca Heavy chain only Antibody is designed to specifically detect Alpaca IgG2c. It is a monoclonal, recombinant, rabbit IgG Fc fused single chain antibody fragment to Alpaca IgG2c coupled to Biotin, and Rabbit Anti-Alpaca IgG2c Biotin Alpaca Heavy chain only Antibody reacts with Alpaca IgG2c selectively, no cross-reactivity with Alpaca IgG1/2b.

The biological family Camelidae comprises camels (one-humped Camelus dromedarius and two-humped Camelus bactrianus), llama (Lama glama and Lama guanicoe), and vicugna (Vicugna vicugna and Vicugna pacos). Camelidae contain two kinds of IgG in serum: conventional antibodies (IgG1) containing two light chains and two heavy chains (composed of the VH, CH1, hinge, and CH2 and CH3 domains) and two types of homodimeric heavy-chain antibodies (HCAbs), IgG2 and IgG3, which comprise only H chains: each H chain contains a VHH, hinge, and CH2 and CH3 domains. The smallest intact functional antigen-binding fragment of HCAbs is the single-domain VHH, also known as a nanobody(Nb). Alpaca is also called Vicugna pacos. Alpaca IgG contains IgG1a, IgG1b, IgG2b, IgG2c and IgG3.

Immunogen

Recombinant Alpaca single variable domain secondary nano antibody

Biochem/physiol Actions

Alpaca IgG2c
Isotype: Rabbit IgG

Packaging

100 μg in Glass bottle

Physical form

Lyophilized from 10mM PBS (pH 7.5), 0.05% sucrose, 0.1% trehalose

Preparation Note

Add 100uL dH2O or 100uL dH2O with 50% Glycerol to a final concentration of 1mg/mL
After reconstitution, store at –20 °C(Avoid freeze / thaw cycles). Stable for 12 months at -20°C

Other Notes

No cross-reactivity with Alpaca IgG1/2b


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

13 - Non Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library