Skip to Content
MilliporeSigma

SBP3387

TAB1 Blocking Peptide for PRS3387

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Form:
liquid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic

form

liquid

species reactivity

human, mouse

concentration

200 μg/mL

shipped in

wet ice

storage temp.

−20°C

Immunogen

13 amino acids near the center of human TAB1.

Application

Used as a blocking peptide in immunoblotting applications.

Other Notes

This peptide blocks the action of antibody PRS3387.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Storage Class

10 - Combustible liquids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library