Skip to Content
MilliporeSigma

SBR00078

Dasabuvir

≥95% (HPLC), powder

Synonym(s):

ABT-333

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C26H27N3O5S
CAS Number:
Molecular Weight:
493.57
UNSPSC Code:
51101500
NACRES:
NA.41
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

powder

antibiotic activity spectrum

viruses

mode of action

enzyme | inhibits

shipped in

ambient

storage temp.

2-8°C

SMILES string

CC(C)(C)C1=CC(=CC(=C1OC)C2=CC3=C(C=C2)C=C(C=C3)NS(=O)(=O)C)N4C=CC(=O)NC4=O

InChI

1S/C26H27N3O5S/c1-26(2,3)22-15-20(29-11-10-23(30)27-25(29)31)14-21(24(22)34-4)18-7-6-17-13-19(28-35(5,32)33)9-8-16(17)12-18/h6-15,28H,1-5H3,(H,27,30,31)

InChI key

NBRBXGKOEOGLOI-UHFFFAOYSA-N

General description

Dasabuvir as a non-nucleoside NS5B polymerase inhibitor which binds the enzyme outside its active site producing a conformational change that inhibits its activity

Biochem/physiol Actions

Dasabuvir is a nonnucleoside inhibitor of the RNA-dependent RNA polymerase encoded by the HCV NS5B gene, inhibits recombinant NS5B polymerases derived from HCV genotype 1a and 1b clinical isolates


Hazard Codes

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library