Skip to Content
MilliporeSigma

SBR00082

Isepamicin Sulfate

≥68%, powder

Synonym(s):

Isepamicin monosulfate, Isepamicin sulfate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C22H45N5O16S
CAS Number:
Molecular Weight:
667.68
UNSPSC Code:
51101500
NACRES:
NA.41
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥68%

form

powder

color

white to off-white

antibiotic activity spectrum

Gram-negative bacteria, Gram-positive bacteria

application(s)

advanced drug delivery

mode of action

protein synthesis | inhibits

storage temp.

2-8°C

SMILES string

C[C@@]1(CO[C@@H]([C@@H]([C@H]1NC)O)O[C@H]2[C@@H](C[C@@H]([C@H]([C@@H]2O)O[C@@H]3[C@@H]([C@H]([C@@H]([C@H](O3)CN)O)O)O)N)NC(=O)[C@H](CN)O)O.OS(=O)(=O)O

InChI

InChI=1S/C22H43N5O12.H2O4S/c1-22(35)6-36-20(15(33)18(22)26-2)39-17-8(27-19(34)9(28)4-23)3-7(25)16(14(17)32)38-21-13(31)12(30)11(29)10(5-24)37-21;1-5(2,3)4/h7-18,20-21,26,28-33,35H,3-6,23-25H2,1-2H3,(H,27,34);(H2,1,2,3,4)/t7-,8+,9-,10+,11+,12-,13+,14-,15+,16+,17-,18+,20+,21+,22-;/m0./s1

InChI key

DDXRHRXGIWOVDQ-MGAUJLSLSA-N

General description

Isepamicin Sulfate is a semisynthetic aminoglycoside antibiotic derived from gentamicin with a spectrum of activity against Gram-positive bacteria, Gram-negative bacteria, and fungi.

Biochem/physiol Actions

Sepamicin Sulfate inhibits bacterial protein synthesis by binding to the bacterial 30S ribosomal subunit, leading to the misreading of genetic information and inhibition of protein synthesis, and ultimately, bacterial cell death.


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library