Skip to Content
MilliporeSigma

SBR00087

Rifamycin S

≥96% (HPLC), powder

Synonym(s):

1,4-Dideoxy-1,4-dioxo-rifamycin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C37H45NO12
CAS Number:
Molecular Weight:
695.75
UNSPSC Code:
51101500
NACRES:
NA.41
eCl@ss:
32035490
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥96% (HPLC)

form

powder

color

yellow-orange

antibiotic activity spectrum

Gram-positive bacteria

application(s)

advanced drug delivery

mode of action

DNA synthesis | inhibits

storage temp.

−20°C

SMILES string

C[C@H]1/C=C/C=C(\C(=O)NC2=CC(=O)C3=C(C2=O)C(=C(C4=C3C(=O)[C@](O4)(O/C=C/[C@@H]([C@H]([C@H]([C@@H]([C@@H]([C@@H]([C@H]1O)C)O)C)OC(=O)C)C)OC)C)C)O)/C

InChI

1S/C37H45NO12/c1-16-11-10-12-17(2)36(46)38-23-15-24(40)26-27(32(23)44)31(43)21(6)34-28(26)35(45)37(8,50-34)48-14-13-25(47-9)18(3)33(49-22(7)39)20(5)30(42)19(4)29(16)41/h10-16,18-20,25,29-30,33,41-43H,1-9H3,(H,38,46)/b11-10+,14-13+,17-12-/t16-,18+,19+,20+,25-,29-,30+,33+,37-/m0/s1

InChI key

BTVYFIMKUHNOBZ-ODRIEIDWSA-N

General description

Rifamycin S is a chemical compound that belongs to the class of antibiotics known as rifamycins. It is a natural product produced by the bacterium Amycolatopsis mediterranei and is used primarily as a reference standard for the detection and quantification of rifamycins in various samples. Rifamycin S is chemically related to other rifamycin antibiotics, including rifampicin.

Application

However, it has been used as a precursor in the synthesis of other rifamycin antibiotics, and it has also been used in research studies to investigate the mechanism of action and resistance of rifamycin antibiotics.

Biochem/physiol Actions

Rifamycin S inhibitions bacterial RNA synthesis. Rifamycin S works by binding to a specific subunit of the bacterial RNA polymerase enzyme, which is necessary for the initiation of bacterial RNA synthesis. Binding of Rifamycin S to the RNA polymerase enzyme inhibits its activity, thereby preventing the transcription of bacterial genes into RNA molecules that are essential for bacterial survival. This leads to the inhibition of bacterial growth and ultimately, bacterial death. Rifamycin S has a broad-spectrum activity against different bacterial species.


Still not finding the right product?

Explore all of our products under Rifamycin S


Pictograms

Health hazard

Signal Word

Warning

Hazard Codes

Precautionary Statements

Hazard Classifications

STOT RE 2 Oral

Target Organs

Kidney

Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library