Skip to Content
MilliporeSigma

SBR00103

Acetylspiramycin

>1200 U/mg

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C45H76N2O15
CAS Number:
Molecular Weight:
885.09
UNSPSC Code:
51101500
NACRES:
NA.21
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

>1200 U/mg

form

powder or solid

storage condition

(close container well)

color

white to off-white, light yellow

mp

131 °C

antibiotic activity spectrum

Gram-positive bacteria

application(s)

advanced drug delivery

mode of action

protein synthesis | inhibits

storage temp.

-10 to -25°C

SMILES string

O=CCC1CC(C)C(OC2OC(C)C(N(C)C)CC2)C=CC=CCC(OC(=O)CC(OC(=O)C)C(OC)C1OC3OC(C)C(OC4OC(C)C(O)C(O)(C)C4)C(N(C)C)C3O)C

InChI

InChI=1S/C45H76N2O15/c1-25-22-31(20-21-48)41(62-44-39(51)38(47(10)11)40(28(4)58-44)61-37-24-45(7,53)43(52)29(5)57-37)42(54-12)34(59-30(6)49)23-35(50)55-26(2)16-14-13-15-17-33(25)60-36-19-18-32(46(8)9)27(3)56-36/h13-15,17,21,25-29,31-34,36-44,51-53H,16,18-20,22-24H2,1-12H3/b14-13+,17-15+/t25-,26-,27-,28-,29+,31+,32+,33+,34-,36+,37+,38-,39-,40-,41+,42+,43+,44+,45-/m1/s1

InChI key

ZPCCSZFPOXBNDL-ZSTSFXQOSA-N

General description

Acetylspiramycin is an antibiotic belonging to the class of macrolides. It is derived from the natural antibiotic spiramycin, which is produced by Streptomyces ambofaciens bacteria.

Biochem/physiol Actions

Mode of Action:
Acetylspiramycin inhibits protein synthesis in susceptible bacteria by binding to the 50S ribosomal subunit of the bacterial ribosome.

Antimicrobial Spectrum:
Active against gram-positive bacteria like taphylococci and streptococci.

Features and Benefits

High quality antibiotic suitable for mulitple research applications Commonly used in Cell Biology and Biochemical applications


Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

Storage Class

11 - Combustible Solids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library