Skip to Content
MilliporeSigma

SML3247

UCSF648 hydrochloride

≥98% (HPLC)

Synonym(s):

5A6-48 hydrochloride, 5A6-78 control hydrochloride, N,N-Dimethyl-2-(quinolin-5-ylmethylamino)ethanesulfonamide hydrochloride, N,N-Dimethyl-2-[(5-quinolinylmethyl)amino]-ethanesulfonamide hydrochloride, UCSF678 control hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.77
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

UCSF648 (5A6-48) is a UCSF678 (5A6-78) analog with no measurable affinity toward any of the 5-HT subtypes. UCSF648 is recommended as a control compound when using UCSF678 for probing 5-HT5A-dependent activities in cultures and in vivo.
UCSF678 (5A6-78) control analog with no measurable affinity toward any of the 5-HT subtypes.


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library