Skip to Content
MilliporeSigma

SML3273

XAF-1407

≥98% (HPLC)

Synonym(s):

(3-methyl-1-[5-phenyl-4-[4-(2-pyrrolidin-1-ylethoxymethyl)-1-piperidyl]thieno[2,3-d]pyrimidin-6-yl]azetidin-3-ol)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C28H37N5O2S
Molecular Weight:
507.69
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

OC1(CN(C1)C2=C(C3=C(N=CN=C3S2)N4CCC(CC4)COCCN5CCCC5)C6=CC=CC=C6)C

InChI

1S/C28H37N5O2S/c1-28(34)18-33(19-28)27-23(22-7-3-2-4-8-22)24-25(29-20-30-26(24)36-27)32-13-9-21(10-14-32)17-35-16-15-31-11-5-6-12-31/h2-4,7-8,20-21,34H,5-6,9-19H2,1H3

InChI key

AVTFTJSBFXQTLR-UHFFFAOYSA-N

Biochem/physiol Actions

Potent and highly selective inhibitor of the acetylcholine-activated inward rectifier potassium current (IKACh) that exhibits an antiarrhythmic effect
XAF-1407 is a potent and highly selective inhibitor of the acetylcholine-activated inward rectifier potassium current (IKACh) that exhibits an antiarrhythmic effect in a goat and horse models of atrial fibrillation. XAF-1407 potently and selectively inhibits ion channels formed by Kir3.1/3.4 heterotetramers and Kir3.4 homotetramers that carry the IKACh current (IC50 of 1.1 and 3.2 nM respectively).


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library