Skip to Content
MilliporeSigma

SML3283

T-0080

≥98% (HPLC)

Synonym(s):

3-(2-Hydroxy-5-methylphenyl)-5-(tetrahydro-2H-pyran-4-yl)-4-(4-(trifluoromethyl)phenyl)-4,5-dihydropyrrolo[3,4-c]pyrazol-6(2H)-one, T 0080, T0080

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C24H22F3N3O3
Molecular Weight:
457.44
UNSPSC Code:
51111800
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C1N(C2CCOCC2)C(C3=CC=C(C(F)(F)F)C=C3)C4=C(C5=C(O)C=CC(C)=C5)NN=C41

InChI

1S/C24H22F3N3O3/c1-13-2-7-18(31)17(12-13)20-19-21(29-28-20)23(32)30(16-8-10-33-11-9-16)22(19)14-3-5-15(6-4-14)24(25,26)27/h2-7,12,16,22,31H,8-11H2,1H3,(H,28,29)

InChI key

NLNJUYHGBAJDBQ-UHFFFAOYSA-N

Biochem/physiol Actions

Blood-brain barrier (BBB)-permeable formyl peptide receptor 1 (FPR1) antagonist with therapeutic efficacy in murine intracerebral hemorrhage (ICH) models in vivo.
T-0080 is a blood-brain barrier (BBB)-permeable formyl peptide receptor 1 (FPR1) antagonist that effectively inhibits fMLF-dependent response in human and murine neutrophils cultures (10 nM fMLF-induced human neutrophil Ca2+ response IC50 = 0.85 μM) and attenuates brain edema and improved neurological outcomes in murine intracerebral hemorrhage (ICH) models in vivo (5 mg/kg via daily i.p.) with no short-term adverse effects (up to 2 g/kg p.o.).


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library