Skip to Content
MilliporeSigma

SML3289

Synucleozid dihydrochloride

≥98% (HPLC)

Synonym(s):

2-(4-(4-Carbamimidoylphenylamino)phenyl)-1H-indole-6-carboximidamide, dihydrochloride, 2-[4-(4-Carbamimidoylanilino)phenyl]-1H-indole-6-carboximidamide, dihydrochloride, 2-[4-[[4-(Aminoiminomethyl)phenyl]amino]phenyl]-1H-indole-6-carboximidamide, dihydrochloride, NSC 377363 2HCl, NSC-377363 2HCl, NSC377363 2HCl

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C22H20N6 · 2HCl
Molecular Weight:
441.36
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

, White to very dark grey

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

[nH]1c2c(cc1c3ccc(cc3)Nc4ccc(cc4)C(=N)N)ccc(c2)C(=N)N

InChI

1S/C22H20N6/c23-21(24)14-5-9-18(10-6-14)27-17-7-3-13(4-8-17)19-11-15-1-2-16(22(25)26)12-20(15)28-19/h1-12,27-28H,(H3,23,24)(H3,25,26)

InChI key

TZRXNWCLKYWDNH-UHFFFAOYSA-N

Biochem/physiol Actions

Selective α-synuclein (α-syn) translation inhibitor with cytoprotective efficacy against preformed α-syn fibrils (PFFs) in SH-SY5Y neuronal cultures.
Synucleozid is a selective α-synuclein (α-syn) translation inhibitor (46% & 69% α-syn downregulation post 48 h, respectively, with 0.5 or 1 μM Synucleozid) with cytoprotective efficacy against preformed α-synuclein fibrils (PFFs) in SH-SY5Y neuronal cultures (56% protection; 0.5 μM Synucleozid added 24 h prior to 48 h 50 μg/mL PFFs exposure). Synucleozid directly and selectively targets α-syn (SNCA) mRNA 5′ UTR at the iron-responsive element (IRE) site, effectively interfering with the polysomes assembly onto SNCA mRNA.


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library