Skip to Content
MilliporeSigma

SML3320

KRA-533

≥98% (HPLC)

Synonym(s):

4-(4-(2-Bromoacetamido)butyl)benzoic acid, 4-[4-[(Bromoacetyl)amino]butyl]benzoic acid, KRAS Agonist-533, NSC 112533, NSC112533, p-[4-(2-Bromoacetamido)butyl]benzoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C13H16BrNO3
CAS Number:
Molecular Weight:
314.18
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(O)C1=CC=C(CCCCNC(CBr)=O)C=C1

InChI

1S/C13H16BrNO3/c14-9-12(16)15-8-2-1-3-10-4-6-11(7-5-10)13(17)18/h4-7H,1-3,8-9H2,(H,15,16)(H,17,18)

InChI key

WZQJLTLURXNPQG-UHFFFAOYSA-N

Biochem/physiol Actions

KRA-333 is a potent and selective KRAS agonist that binds directly to the GTP/GDP binding pocket of KRAS preventing the cleavage of GTP into GDP and caucusing accumulation of activated GTP-KRAS and suppression of cell growth. KRA-333 promotes apoptosis and autophagic cell death of cancer cells and inhibits growth of mutant KRAS lung cancer in xenografts mice model.
Potent and selective KRAS agonist that binds directly to the GTP/GDP binding pocket caucusing accumulation of activated GTP-KRAS and suppression of cell growth


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library