Skip to Content
MilliporeSigma

SML3352

GSK973

≥98% (HPLC)

Synonym(s):

(2S,3S)-N5-((1R,5S,6r)-3-oxabicyclo[3.1.0]hexan-6-yl)-2-(fluoromethyl)-N7-methyl-3-phenyl-2,3-dihydrobenzofuran-5,7-dicarboxamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C23H23FN2O4
CAS Number:
Molecular Weight:
410.44
UNSPSC Code:
12352200
MDL number:
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

Orally available, potent and selective pan-second bromodomain (BD2) inhibitor that effectively inhibits LPS-induced MCP-1 release in human whole blood.



GSK973 is an orally available, potent and selective pan-second bromodomain (BD2) inhibitor (BD2/BD1 pIC50 = 7.5/4.4/BRD2, 7.8/4.5/BRD3, 7.8/4.6/BRD4, 7.4/4.5/BRDT) that effectively inhibits LPS-induced MCP-1 release in human whole blood (pIC50 = 7.3; IC50 = 50 nM).


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library