Skip to Content
MilliporeSigma

SML3353

GSK943

≥98% (HPLC)

Synonym(s):

(2R,3R)-N5-((1R,5S,6r)-3-oxabicyclo[3.1.0]hexan-6-yl)-2-(fluoromethyl)-N7-methyl-3-phenyl-2,3-dihydrobenzofuran-5,7-dicarboxamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C23H23FN2O4
CAS Number:
Molecular Weight:
410.44
UNSPSC Code:
12352200
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CNC(C1=C2C([C@](C3=CC=CC=C3)([H])[C@@H](O2)CF)=CC(C(N[C@@H]4[C@]5([H])[C@@]4([H])COC5)=O)=C1)=O

InChI key

WZQLVEPIBAOOGF-PXTPFGJHSA-N

Biochem/physiol Actions

Inactive (2R,3R) stereoisomer of and the recommended negative control for the potent and selective pan-second bromodomain (BD2) inhibitor GSK973.



GSK943 corresponds to the inactive (2R,3R) stereoisomer of and the recommended negative control for the potent and selective pan-second bromodomain (BD2) inhibitor GSK973.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library