Skip to Content
MilliporeSigma

SML3372

Glutipyran

≥98% (HPLC)

Synonym(s):

rel-N-(3,4-Dimethoxybenzyl)-N-((R)-3-((S)-2,2-dimethyltetrahydro-2H-pyran-4-yl)-3-phenylpropyl)furan-2-carboxamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C30H37NO5
Molecular Weight:
491.62
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

shipped in

dry ice

storage temp.

-10 to -25°C

Biochem/physiol Actions

Broad-spectrum potent and selective glucose transporter (GLUT) inhibitor
Glutipyran is a broad-spectrum potent and selective glucose transporter (GLUT) inhibitor that targets both GLUT1 and GLUT3. Glutipyran inhibits glycolytic activity and cell growth in PANC-1 cells (IC50 = 1.9 µM). It exhibits antitumor activity (12.5 mg/kg, i.v.) once daily in MIA PaCa-2 xenograft mouse model.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library