Skip to Content
MilliporeSigma

SML3401

NVP-LMD194

≥98% (HPLC)

Synonym(s):

N-[(6-Amino-3-pyridinyl)methyl]-N′-[4-[(1,3-dihydro-1,3-dioxo-2H-isoindol-2-yl)methyl]phenyl]urea

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C22H19N5O3
CAS Number:
Molecular Weight:
401.42
NACRES:
NA.77
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

2-8°C

Biochem/physiol Actions

Cellularly active, potent, and selective inhibitor of NAMPT (nicotinamide phosphoribosyltransferase)
NVP-LMD194 is a cellularly active, potent, and selective inhibitor of NAMPT (nicotinamide phosphoribosyltransferase). NVP-LMD194 decreases the viability of A2780 and CORL23 cells.


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library