Skip to Content
MilliporeSigma

SML3407

5-NIDR

≥98% (HPLC)

Synonym(s):

1-(2-Deoxy-β-D-erythro-pentofuranosyl)-5-nitro-1H-indole, 5-Nitroindole 2′-deoxyribonucleoside

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C13H14N2O5
CAS Number:
Molecular Weight:
278.26
NACRES:
NA.77
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

packaging

pkg of 4 vials (Amber)

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C



Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library