Skip to Content
MilliporeSigma

SML3418

Xanthatin

≥98% (HPLC)

Synonym(s):

(-)-Xanthatin, (3aR,​7S,​8aS)​-3,​3a,​4,​7,​8,​8a-​hexahydro-​7-​methyl-​3-​methylene-​6-​[(1E)​-​3-​oxo-​1-​buten-​1-​yl]​-​2H-​cyclohepta[b]​furan-​2-​one, CID 5281511

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C15H18O3
CAS Number:
Molecular Weight:
246.30
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

O1[C@@H]2[C@H](CC=C([C@H](C2)C)\C=C\C(=O)C)C(=C)C1=O

InChI

1S/C15H18O3/c1-9-8-14-13(11(3)15(17)18-14)7-6-12(9)5-4-10(2)16/h4-6,9,13-14H,3,7-8H2,1-2H3/b5-4+/t9-,13+,14-/m0/s1

InChI key

RBRPTFMVULVGIC-ZTIIIDENSA-N

Biochem/physiol Actions

Potent anticancer agent that induces cell cycle arrest at the G2/M phase.

Xanthatin, a sesquiterpene lactone isolated from Xanthium plants (Asteraceae), is a potent anticancer agent that induces cell cycle arrest at the G2/M phase in hepatocellular carcinoma and colon cancer cells. Xanthatin induces apoptosis in numerous cancer cell lines including colon, breast, lung, cervix, and skin cancers, while apparently spearing human normal LO2 hepatocytes. It appears to activate endoplasmic reticulum stress through activation of the PERK/eIF-2α/ATF4 axis in hepatocellular carcinoma cells. Also, xanthatin exhibit potent phytotoxic activity, which suppressed root growth of receiver plants.


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library