Skip to Content
MilliporeSigma

SML3428

GLPG2938

≥97% (HPLC)

Synonym(s):

1-[2-Ethoxy-6-(trifluoromethyl)-4-pyridyl]-3-[[5-methyl-6-[1-methyl-3-(trifluoromethyl)pyrazol-4-yl]pyridazin-3-yl]methyl]urea, GLPG 2938, GLPG-2938, N-[2-Ethoxy-6-(trifluoromethyl)-4-pyridinyl]-N′-[[5-methyl-6-[1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]-3-pyridazinyl]methyl]urea

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H19F6N7O2
CAS Number:
Molecular Weight:
503.40
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥97% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥97% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

O=C(NCC1=CC(C)=C(C2=CN(C)N=C2C(F)(F)F)N=N1)NC3=CC(C(F)(F)F)=NC(OCC)=C3

InChI

1S/C20H19F6N7O2/c1-4-35-15-7-11(6-14(29-15)19(21,22)23)28-18(34)27-8-12-5-10(2)16(31-30-12)13-9-33(3)32-17(13)20(24,25)26/h5-7,9H,4,8H2,1-3H3,(H2,27,28,29,34)

InChI key

MGJMUVKYINFAQC-UHFFFAOYSA-N

Biochem/physiol Actions

GLPG2938 is an orally active, potent and selective S1P2 receptor (S1PR2) antagonist (GTPγS induction IC50 = 29 nM; [S1P] = EC80) with no activity against other S1P receptors. GLPG2938 inhibits sphingosine 1-phosphate (S1P)-induced cellular responses in cultures (Ca2+ flux IC50 = 8.8 nM & IL8 release IC50 = 3.8 nM; [S1P] = EC80) and exhibits good efficacy in a 14-day model of bleomycin-induced pulmonary fibrosis in mice in vivo (1, 3, and 10 mg/kg b.i.d. p.o.) with good pharmacokinetics.
Orally active, potent and selective S1P2 subtype-selective sphingosine-1-phosphate (S1P) receptor antagonist with in vitro and in vivo efficacy.


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library