Skip to Content
MilliporeSigma

SML3436

S-3-Carboxypropyl-L-cysteine

≥98% (HPLC)

Synonym(s):

(R)​-​4-​((2-​Amino-​2-​carboxyethyl)​thio)​butanoic acid, S-​(3-​Carboxypropyl)​-​L-​cysteine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C7H13NO4S
CAS Number:
Molecular Weight:
207.25
UNSPSC Code:
12352209
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

2 mg/mL, clear (0.1N HCl

Warmed)

storage temp.

-10 to -25°C

SMILES string

S(C[C@H](N)C(=O)O)CCCC(=O)O

InChI

1S/C7H13NO4S/c8-5(7(11)12)4-13-3-1-2-6(9)10/h5H,1-4,8H2,(H,9,10)(H,11,12)/t5-/m0/s1

InChI key

WNFNRNDFHINZLV-YFKPBYRVSA-N

Biochem/physiol Actions

Potent and selective reversible inhibitor of Cystathionine γ-lyase.
S-3-Carboxypropyl-L-cysteine is a potent and selective reversible inhibitor of Cystathionine γ-lyase that inhibits both the cystathionine and cysteine cleavage reactions. S-3-Carboxypropyl-L-cysteine inhibits the transsulfuration pathway and H2S production in cells. It does not inhibit cystathionine β-synthase and mercaptopyruvate sulfurtransferase.


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library