Skip to Content
MilliporeSigma

SML3506

PD 173212

≥95% (HPLC)

Synonym(s):

(S)-N-((S)-3-(4-(Benzyloxy)phenyl)-1-(tert-butylamino)-1-oxopropan-2-yl)-2-((4-tert-butylbenzyl)(methyl)amino)-4-methylpentanamide, N‐[[4‐(1,1‐Dimethylethyl)phenyl]methyl]‐N‐methyl‐L‐leucyl‐N‐(1,1‐dimethylethyl)‐O‐(phenylmethyl)‐L‐tyrosinamide, PD-173212

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C38H53N3O3
CAS Number:
Molecular Weight:
599.85
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
eCl@ss:
32160703
Assay:
≥95% (HPLC)
Form:
(Powder, lyophilized powder, or film)
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

(Powder, lyophilized powder, or film)

storage condition

desiccated

color

white to off-white

storage temp.

-10 to -25°C

SMILES string

O=C([C@@H](NC([C@@H](N(C)CC1=CC=C(C=C1)C(C)(C)C)CC(C)C)=O)CC2=CC=C(OCC3=CC=CC=C3)C=C2)NC(C)(C)C

InChI

1S/C38H53N3O3/c1-27(2)23-34(41(9)25-29-15-19-31(20-16-29)37(3,4)5)36(43)39-33(35(42)40-38(6,7)8)24-28-17-21-32(22-18-28)44-26-30-13-11-10-12-14-30/h10-22,27,33-34H,23-26H2,1-9H3,(H,39,43)(H,40,42)/t33-,34-/m0/s1

InChI key

GCDHMGROXQUFNR-HEVIKAOCSA-N

Biochem/physiol Actions

PD 173212 is a selective N-type (Cav2.2) voltage sensitive calcium channel (VSCC) blocker that potently inhibits VSCC-mediated calcium influx in IMR-32 human neuroblastoma cells (IC50 = 36 nM vs. 8%/Na+ channels & 10% K+ channels blockage at 1 μM, L-type Ca+2 channels IC50 >10 μM) with in vivo efficacy in a murine audiogenic zeisure model (80% tonic zeisure protection at 30 mg/kg i.v.).
Selective N-type voltage-gated calcium channel (VSCC) blocker with in vivo efficacy in a murine audiogenic seizure model.


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library