Skip to Content
MilliporeSigma

SML3512

CP-681301

≥98% (HPLC)

Synonym(s):

CP 681301, CP681301, N-(3-Cyclobutyl-1H-pyrazol-5-yl)-2-(4-(dimethylamino)phenyl)acetamide, N-(5-Cyclobutyl-1H-pyrazol-3-yl)-2-(4-dimethylaminophenyl)acetamide, N-(5-Cyclobutyl-2H-pyrazol-3-yl)-2-(4-dimethylaminophenyl)acetamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C17H22N4O
CAS Number:
Molecular Weight:
298.38
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

O=C(CC1=CC=C(C=C1)N(C)C)NC2=CC(C3CCC3)=NN2

InChI

1S/C17H22N4O/c1-21(2)14-8-6-12(7-9-14)10-17(22)18-16-11-15(19-20-16)13-4-3-5-13/h6-9,11,13H,3-5,10H2,1-2H3,(H2,18,19,20,22)

InChI key

PGTNTDKBRSQDNM-UHFFFAOYSA-N

Biochem/physiol Actions

ATP-competitive, potent and selective inhibitor against cyclin-dependent kinase Cdk5 both in vitro and in vivo.
CP-681301 is a potent, ATP-competitive cyclin-dependent kinase Cdk5 inhibitor (Ki = 13.7 nM; 2.8-/40-fold selectivity over Cdk2/GSK-3β) that blocks Cdk5 substrates phosphorylation in cultures (tau pS235 IC50 = 513 nM in CHO p25/CDK5/tau transfectants; IC50 ~500 nM against 48-h 10 µM oligomeric Aβ42-stimulated APP pT668 in murine embryonic neurons) and reverses the upregulated tau pS235 level in Cdk5 activator p25-overexpressing transgenic neonatal mice in vivo (5.8 mg/kg s.c.; 6 hrs). CP-681301 selectively inhibits Cdk5-dependent proliferation of human medullary thyroid carcinoma (MTC), but not normal human fibroblasts in cultures (5 µM, 48 hrs).


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library