Skip to Content
MilliporeSigma

SML3538

CKJB71

≥98% (HPLC)

Synonym(s):

5-(4-Methylpiperazine-1-carbonyl)-7,10-dioxa-13,17,18,21-tetraazatetracyclo[12.5.2.1²,⁶.0¹⁷,²⁰]docosa-1(20),2,4,6(22),14(21),15,18-heptaene, CK156 inactive control, CK156 negative control

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C22H26N6O3
Molecular Weight:
422.48
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(N1CCN(C)CC1)C(C=C2)=C(OCCOCCNC3=N4)C=C2C5=C4N(C=C3)N=C5

InChI

1S/C22H26N6O3/c1-26-7-9-27(10-8-26)22(29)17-3-2-16-14-19(17)31-13-12-30-11-5-23-20-4-6-28-21(25-20)18(16)15-24-28/h2-4,6,14-15H,5,7-13H2,1H3,(H,23,25)

InChI key

DDXOMWWURGFLKK-UHFFFAOYSA-N

Biochem/physiol Actions

CKJB71 is an inactive analog of and negative control (IC50 = 20 μM and >45 μM using cell lysate or by inact cell NanoBRET) for the potent and selective DRAK1 (STK17A) inhibitor CK156.
Inactive analog of and negative control for the potent and selective DRAK1 (STK17A) inhibitor CK156.


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library