Skip to Content
MilliporeSigma

SML3550

PAIR1

≥98% (HPLC)

Synonym(s):

(S)-2,2,2-trifluoro-N-(4-((3-(2-(piperidin-3-ylamino)pyrimidin-4-yl)pyridin-2-yl)oxy)naphthalen-1-yl)ethane-1-sulfonamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C26H25F3N6O3S
CAS Number:
Molecular Weight:
558.58
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=S(NC(C=C1)=C2C(C=CC=C2)=C1OC3=NC=CC=C3C4=CC=NC(N[C@@H]5CNCCC5)=N4)(CC(F)(F)F)=O

InChI

1S/C26H25F3N6O3S/c27-26(28,29)16-39(36,37)35-22-9-10-23(19-7-2-1-6-18(19)22)38-24-20(8-4-13-31-24)21-11-14-32-25(34-21)33-17-5-3-12-30-15-17/h1-2,4,6-11,13-14,17,30,35H,3,5,12,15-16H2,(H,32,33,34)/t17-/m0/s1

InChI key

LVBYPSVRFGNRPW-KRWDZBQOSA-N

Biochem/physiol Actions

Cell penetrant, potent and selective ATP-competitive partial antagonist of the IRE1α RNase activity, while affording quantitative kinase activity inhibition
PAIR1 is a cell penetrant, potent and selective ATP-competitive partial antagonist of the IRE1α (endoplasmic reticulum to nucleus signaling 1) RNase activity, while affording quantitative kinase activity inhibition. PAIR1 partially antagonize the IRE1α RNase at full occupancy of the kinase. In β-cells PAIR1 allows adaptive splicing of Xbp1 mRNA while suppressing destructive ER mRNA endonucleolytic decay and apoptosis.


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library