Skip to Content
MilliporeSigma

SML3560

MU380

≥98% (HPLC)

Synonym(s):

(R)-6-bromo-5-(piperidin-3-yl)-3-(1-(trifluoromethyl)-1H-pyrazol-4-yl)pyrazolo[1,5-a]pyrimidin-7-amine; 6-Bromo-5-(3R)-3-piperidinyl-3-[1-(trifluoromethyl)-1H-pyrazol-4-yl]-pyrazolo[1,5-a]pyrimidin-7-amine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C15H15BrF3N7
CAS Number:
Molecular Weight:
430.23
UNSPSC Code:
12352200
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

NC1=C(C([C@@]2([H])CNCCC2)=NC3=C(C=NN13)C4=CN(N=C4)C(F)(F)F)Br

Biochem/physiol Actions

MU380 is a metabolically stable analog of CHK1 inhibitor SCH900776 (MK8776) does not undergo in vivo N-dealkylation. MU380 is a potent and selective CHK1 inhibitor, which potently sensitizes multiple cancer cell lines to DNA antimetabolites including hydroxyurea, cytarabine and gemcitabine.
Metabolically stable, potent and selective CHK1 inhibitor


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library