Skip to Content
MilliporeSigma

SML3567

EN450

≥98% (HPLC)

Synonym(s):

N-(2-Chloro-5-(N,N-dimethylsulfamoyl)phenyl)acrylamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C11H13ClN2O3S
CAS Number:
Molecular Weight:
288.75
UNSPSC Code:
12352200
NACRES:
NA.21
eCl@ss:
32150313
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(NC1=CC(S(=O)(N(C)C)=O)=CC=C1Cl)C=C

InChI key

OXFXDOLAQBMOPH-UHFFFAOYSA-N

Biochem/physiol Actions

Cysteine reactive UBE2D/NFKB1 molecular glue degrader; NFKB1 molecular glue degrader
EN450 is a molecular glue degraders of the oncogenic transcription factor NFKB1 that induces ternary complex formation between ubiquitin-conjugating enzyme E2D1 (UBE2D1) and NFKB1. EN450 covalently modifies an allosteric C111 in UBE2D1 and significantly reduces NFKB1 p105 levels. It impairs leukemia cell viability in a NEDDylation and proteasome-dependent manner at 50 µM.


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library