Skip to Content
MilliporeSigma

SML3570

M4K2234NC

≥98% (HPLC)

Synonym(s):

(2-Fluoro-4-(5-(4-(4-isopropylpiperazin-1-yl)phenyl)-4-methylpyridin-3-yl)-6-methoxyphenyl)(pyrrolidin-1-yl), M4K2234 inactive control, M4K2234 negative control

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C31H37FN4O2
Molecular Weight:
516.65
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

2-8°C

SMILES string

CC(C)N1CCN(CC1)C2=CC=C(C3=C(C)C(C4=CC(OC)=C(C(N5CCCC5)=O)C(F)=C4)=CN=C3)C=C2

InChI key

WFJOMSKCDAOJBT-UHFFFAOYSA-N

Biochem/physiol Actions

Inactive analog of and negative control for the potent and selective TGF-β type I receptors ALK1/ACVRL1 & ALK2/ACVR1 inhibitor M4K2234.
M4K2234NC is an inactive analog of and the recommended negative control for the potent and selective TGF-β type I receptors ALK1/ACVRL1 & ALK2/ACVR1 inhibitor M4K2234 (ALK1/2 IC50 >10 μM/ 5.69 μM by cellular NanoBRET assays; IC50 = 83 nM/13 nM using M4K2234).


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library