Skip to Content
MilliporeSigma

SML3580

Rapa*-3, mixture of E and Z isomers

≥98% (HPLC)

Synonym(s):

27-(O-Ethyloxime)-rapamycin, Rapa 3(E/Z), Rapa-3(E/Z), Rapa3(E/Z)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C53H84N2O13
Molecular Weight:
957.24
UNSPSC Code:
12352200
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

O=C1O[C@]([C@@H](C)C[C@H]2CC[C@H](O)[C@@H](OC)C2)([H])CC([C@@H](C)/C=C([C@H](O)[C@H](OC)C([C@@H](C)C[C@@H](C)/C=C/C=C/C=C([C@H](OC)C[C@]3([H])O[C@@](O)([C@@H](C)CC3)C(C(N4CCCC[C@]14[H])=O)=O)\C)=O)\C)=NOCC

Biochem/physiol Actions

Rapa*-3Z is a rapamycin analog that selectively targets FKBP12 with V55G mutation, while exhibiting reduced affinity toward the wild-type FKBP12 (mTOR FRB association induction EC50 = 11.8 nM/V55G vs 639 nM/wt FKBP12). Rapa*-3Z selectively inhibits the growth, the phosphorylation level of mTORC1 downstream pS6K/pS6 and mTORC2 downstream pAkt in 293 transfectants experssing the V55G FKBP12 mutant (mutant cells pS6K/pS6 IC50 = 1.0/1.3 nM vs. 76/121 nM in wt cells). Rapa*-3Z potently inhibits the larval development of transgenic flies that express human FKBP12 V55G (by 70%/75 μM and 100%/100 μM), but not those with wild-type human FKBP12.
Rapamycin analog that selectively inhibits mTOR signaling in FKBP12(V55G) mutant-expressing cells and organisms.


Pictograms

Health hazard

Signal Word

Warning

Hazard Codes

Hazard Classifications

Carc. 2 - Repr. 2

Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library