Skip to Content
MilliporeSigma

SML3606

Nosiheptide

≥97% (HPLC)

Synonym(s):

Multhiomycin, N-[1-(aminocarbonyl)ethenyl]-2-[(11S,14Z,21S,23S,29S)-14-ethylidene-9,10,11,12,13,14,19,20,21,22,23,24,26,33,35,36-hexadecahydro-3,23-dihydroxy-11-[(1R)-1-hydroxyethyl]-31-methyl-9,12,19,24,33,43-hexaoxo-30,32-imino-8,5:18,15:40,37-trinitrilo-21,36-([2,4]-endothiazolomethanimino)-4-thiazolecarboxamide, NSC 307240, RP 9671

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C51H43N13O12S6
CAS Number:
Molecular Weight:
1222.36
UNSPSC Code:
51111800
Assay:
≥97% (HPLC)
Form:
(Powder or Lyophilized powder or film)
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥97% (HPLC)

form

(Powder or Lyophilized powder or film)

storage condition

desiccated

color

white to beige

storage temp.

-10 to -25°C

SMILES string

[s]1c(nc(c1)C(=O)NC(=C)C(=O)N)c2nc3c(cc2O)c4[s]cc(n4)C(=O)NC(C(=O)NC(=CC)c5[s]cc(n5)C(=O)NC6CC(C(=O)OCc7c8c([nH]c(c8C)C(=O)SCC(NC(=O)c%10nc6[s]c%10)c9[s]cc3n9)ccc7)O)C(O)C

InChI

1S/C51H43N13O12S6/c1-5-23-46-60-28(14-79-46)41(70)56-25-10-33(67)50(74)76-11-21-7-6-8-24-34(21)18(2)35(54-24)51(75)82-17-31(57-42(71)29-15-80-47(25)61-29)48-58-26(12-78-48)37-22(45-59-30(13-77-45)43(72)64-36(20(4)65)44(73)55-23)9-32(66)38(63-37)49-62-27(1

InChI key

OQAOHXRUMXWDLQ-UHFFFAOYSA-N

Biochem/physiol Actions

Thiopeptide antibiotic.

Nosiheptide is a thiopeptide antibiotic effective against Gram-positive bacteria with potent activity against methicillin-resistant Staphylococcus aureus (MRSA). It is inactive against most Gram-negative bacteria. Nosiheptide inhibits bacterial ribosomal protein synthesis by binding to the 50S ribosomal subunit blocking the binding region of elongation factor. It is not approved as an antibiotic for human use, but has found veterinary use in feed as an animal growth promoter.

Disclaimer

Hygroscopic


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library