Skip to Content
MilliporeSigma

SML3632

NAQ

≥98% (HPLC)

Synonym(s):

17-Cyclopropylmethyl-3,14b-dihydroxy-4,5a-epoxy-6a-[(3’-isoquinolyl)acetamido]morphinan dihydrochloride, N-[(5α,6α)-17-(Cyclopropylmethyl)-4,5-epoxy-3,14-dihydroxymorphinan-6-yl]-3-isoquinolinecarboxamide dihydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C30H31N3O4·2HCl
CAS Number:
Molecular Weight:
570.51
UNSPSC Code:
12352116
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O[C@]12[C@@]34C5=C(C[C@H]2N(CC4)CC6CC6)C=CC(O)=C5O[C@@]3([H])[C@H](CC1)NC(C7=CC8=C(C=CC=C8)C=N7)=O.[2HCl]

Biochem/physiol Actions

Highly selective and high-affinity mu (μ) opioid receptor (MOR) antagonist in vitro and in vivo.




NAQ is a highly selective and high-affinity mu (μ) opioid receptor (MOR) antagonist with 241- & 48-fold selectivity over the delta (δ) and kappa (κ) opioid receptor, respectively (MOR/DOR/KOR Ki = 0.55/132.5/26.45 nM) and little MOR agonist activity (Emax = 15.83% of that of DAMGO by GTP[γS] binding using membrane from MOR-expressing cells). NAQ effectively antagonizes morphine antinociceptive effects in mice in vivo (warm-water tail immersion test AD50 = 0.45 mg/kg s.c. 5 min prior to 10 mg morphine/kg s.c.; no agonist efficacy up to 100 mg/kg).

Disclaimer

Hygroscopic


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library