Skip to Content
MilliporeSigma

SML3640

GS-6201

≥98% (HPLC)

Synonym(s):

3-Ethyl-3,9-dihydro-1-propyl-8-[1-[[3-(trifluoromethyl)phenyl]methyl]-1H-pyrazol-4-yl]-1H-purine-2,6-dione, 3-ethyl-1-propyl-8-[1-(3-trifluoromethylbenzyl)-1H-pyrazol-4-yl]-3,7-dihydropurine-2,6-dione, CVT 6883, CVT-6883

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C21H21F3N6O2
CAS Number:
Molecular Weight:
446.43
UNSPSC Code:
12352200
NACRES:
NA.21
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

-10 to -25°C

Biochem/physiol Actions

GS-6201 (CVT-6883) is a potent and selective A2B adenosine receptor (A2BAdoR) antagonist. GS-6201 reduces caspase-1 activity in the heart and leads to a more favorable cardiac remodeling in a mouse model of non-reperfused myocardial infarction. Also GS-6201 attenuated vascular remodeling and hypertension in mouse model.
Potent and selective A2B adenosine receptor (A2BAdoR) antagonist


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library