Skip to Content
MilliporeSigma

SML3665

Palosuran

≥98% (HPLC)

Synonym(s):

1-[2-(4-benzyl-4-hydroxy-piperidin-1-yl)-ethyl]-3-(2-methyl-quinolin-4-yl)-urea, CT-058362

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C25H30N4O2
CAS Number:
Molecular Weight:
418.53
UNSPSC Code:
12352200
NACRES:
NA.25
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

N3(CCC(CC3)(O)Cc4ccccc4)CCNC(=O)Nc1c2c(nc(c1)C)cccc2

InChI

1S/C25H30N4O2/c1-19-17-23(21-9-5-6-10-22(21)27-19)28-24(30)26-13-16-29-14-11-25(31,12-15-29)18-20-7-3-2-4-8-20/h2-10,17,31H,11-16,18H2,1H3,(H2,26,27,28,30)

InChI key

WYJCYXOCHXWTHG-UHFFFAOYSA-N

Biochem/physiol Actions

Orally available, potent, and selective noncompetitive urotensin 2 receptor antagonist
Palosuran (ACT-058362) is an orally available, potent, and selective noncompetitive urotensin 2 receptor antagonist. Palosuran decreases serum glucose, cholesterol, and triglyceride levels and increases survival in a rat model of diabetes induced by streptozotocin.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library