Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C15H10F6OS2
CAS Number:
Molecular Weight:
384.36
UNSPSC Code:
12352200
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder
Quality Segment
assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 2 mg/mL, clear (Warmed)
storage temp.
-10 to -25°C
SMILES string
FC(F)(F)SC1=CC=C(COC2=CC=C(C=C2)SC(F)(F)F)C=C1
InChI
1S/C15H10F6OS2/c16-14(17,18)23-12-5-1-10(2-6-12)9-22-11-3-7-13(8-4-11)24-15(19,20)21/h1-8H,9H2
InChI key
OJUUGOMDHJFXJH-UHFFFAOYSA-N
Biochem/physiol Actions
Cell penetrant potent and selective inhibitor of Receptor Protein Tyrosine Phosphatase (RPTP) β/ζ.
MY10 is a cell penetrant potent and selective inhibitor of Receptor Protein Tyrosine Phosphatase (RPTP) β/ζ. It interacts with the cytoplasmatic D1 domain of RPTPβ/ζ. MY10 reduces alcohol intake in rats.
Storage Class
11 - Combustible Solids
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.