Skip to Content
MilliporeSigma

SML3732

BAY-4931

≥98% (HPLC)

Synonym(s):

2-Chloro-N-[2-(4-ethylphenyl)-1,3-benzoxazol-5-yl]-5-nitrobenzamide, BAY 4931, BAY4931

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C22H16ClN3O4
CAS Number:
Molecular Weight:
421.83
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(C1=CC([N+]([O-])=O)=CC=C1Cl)NC2=CC=C3OC(C4=CC=C(C=C4)CC)=NC3=C2

InChI

1S/C22H16ClN3O4/c1-2-13-3-5-14(6-4-13)22-25-19-11-15(7-10-20(19)30-22)24-21(27)17-12-16(26(28)29)8-9-18(17)23/h3-12H,2H2,1H3,(H,24,27)

InChI key

WFEBALYYDASKIV-UHFFFAOYSA-N

Biochem/physiol Actions

BAY-4931 is a potent and selective covalent PPARγ inverse agonist (IC50 = 0.17 nM, max inhibition = 94% by cell-based reporter assays) that induces interactions between the PPARγ ligand-binding domain (LBD) and the corepressor NCOR2 (EC50 = 5.8 nM, Emax = 619% by TR-FRET). BAY-4931 exhibits anti-proliferation efficacy against PPARγ-dependent growth in cultures (UM-UC9 IC50 = 3.4 nM, max inhibition = 79%).
Potent and selective covalent PPARγ inverse agonist with anti-proliferation efficacy against PPARγ-dependent growth in cultures.


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library