Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C22H16BrN3O4
CAS Number:
Molecular Weight:
466.28
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Quality Segment
assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 2 mg/mL, clear
storage temp.
2-8°C
Biochem/physiol Actions
BAY-0069 is a potent and selective covalent PPARγ inverse agonist (IC50 = 0.22 nM, max inhibition = 88% by cell-based reporter assays) that induces interactions between the PPARγ ligand-binding domain (LBD) and the corepressor NCOR2 (EC50 = 9.6 nM, Emax = 620% by TR-FRET). BAY-0069 exhibits anti-proliferation efficacy against PPARγ-dependent growth in cultures (UM-UC9 IC50 = 2.5 nM, max inhibition = 72%).
Potent and selective covalent PPARγ inverse agonist with anti-proliferation efficacy against PPARγ-dependent growth in cultures.
Storage Class
11 - Combustible Solids
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.