Skip to Content
MilliporeSigma

SML3741

GK16S

≥98% (HPLC)

Synonym(s):

(S)-1-Cyano-N-(pent-4-yn-1-yl)pyrrolidine-3-carboxamide (GK16S)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C11H15N3O
Molecular Weight:
205.26
NACRES:
NA.77
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
oil
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

oil

color

yellow

storage temp.

2-8°C

Biochem/physiol Actions

GK16S is a non-toxic, chemogenomic control for GK13S to investigate the specific inhibition of UCHL1. GK16S does not exhibit any appreciable degree of inhibition of UCHL1, but engages all lesser targets of GK13S (such as PARK7, C21orf33, ISOC1 andNIT2). GK13S together with GK16S comprises a set of chemogenomic probes for UCHL1, which is suitable for investigation of cellular functions of UCHL.
Non-toxic, chemogenomic control for GK13S to investigate the specific inhibition of UCHL1


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library