Skip to Content
MilliporeSigma

SML3746

TFCH

≥98% (HPLC)

Synonym(s):

7-Hydrazinyl-4-(trifluoromethyl)-2H-1-benzopyran-2-one

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C10H7F3N2O2
CAS Number:
Molecular Weight:
244.17
UNSPSC Code:
12352200
NACRES:
NA.21
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

, Faint yellow to yellow brown

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

Brain penetrant, photo-stable, hydrazinyl coumarin compound that acts as a two-photon highly selective fluorescent sensor for both malondialdehyde and formaldehyde
TFCH is a photo-stable, hydrazinyl coumarin compound that acts as a two-photon fluorescent sensor for biomolecule carbonyls with low biotoxicity and extraordinary biocompatibility. TFCH is blood-brain barrier permeant, highly selective for both malondialdehyde (MDA) and formaldehyde (FA) with low detection levels (0.13 & 0.11 uM), respectively. It is shown to simultaneously and separately detect and image MDA and FA (0.11 uM) levels in living brains (0.37 mg/kg, i.p.) and in PC12 cells by two-photon microscopy. TFCH efficiently detects modest forms of renal injury with much greater sensitivity than standard assays for nephrotoxins. For MDA: Ex 365 & 800 nm, Em 440 nm; For FA: Ex 365 & 800 nm, Em 510 nm.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library