Skip to Content
MilliporeSigma

SML3798

Iptakalim hydrochloride

≥98% (HPLC)

Synonym(s):

2,3-dimethyl-N-(1-methylethyl)-2-butanamine hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C9H21N·HCl
CAS Number:
Molecular Weight:
179.73
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

[Cl-].[N+H2](C(C(C)C)(C)C)C(C)C

InChI

1S/C9H21N.ClH/c1-7(2)9(5,6)10-8(3)4;/h7-8,10H,1-6H3;1H

InChI key

RXNJQAYLNOXYKW-UHFFFAOYSA-N

Biochem/physiol Actions

Brain blood barrier penetrant ATP-sensitive potassium channel (KATP) opener that attenuates hypoxia-induced pulmonary arterial hypertension in rats; K(ir) 6.1/SUR2B activator.
Iptakalim hydrochloride is a brain blood barrier penetrant ATP-sensitive potassium channel (KATP) opener that attenuates hypoxia-induced pulmonary arterial hypertension in rats. Iptakalim hydrochloride is a selective antagonist of α4β2-containing nicotinic acetylcholine receptor (nAChR).

Disclaimer

Hygroscopic


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library