Skip to Content
MilliporeSigma

SML3819

UNC-CA157

≥98% (HPLC)

Synonym(s):

4-((3,4,5-Trimethoxyphenyl)amino)quinoline-7-carbonitrile, 4-[(3,4,5-Trimethoxyphenyl)amino]-7-quinolinecarbonitrile

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C19H17N3O3
CAS Number:
Molecular Weight:
335.36
NACRES:
NA.77
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

Cell penetrant, selective and potent inhibitor of archetypal UbiB protein COQ8.
UNC-CA157 is a cell penetrant, selective and potent inhibitor of COQ8 the member of archetypal UbiB family of atypical kinases (protein kinase-like (PKL) family). UNC-CA157 inhibits CoQ production in COQ8BKO cells.


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library