Skip to Content
MilliporeSigma

SML3857

Etamicastat

≥98% (HPLC)

Synonym(s):

(R)-5-(2-Aminoethyl)-1-(6,8-difluorochroman-3-yl)-1,3-dihydroimidazole-2-thione, (R)-5-(2-Aminoethyl)-1-(6,8-difluorochroman-3-yl)-1H-imidazole-2(3H)-thione, 5-​(2-​Aminoethyl)​-​1-​[(3R)​-​6,​8-​difluoro-​3,​4-​dihydro-​2H-​1-​benzopyran-​3-​yl]​-​1,​3-​dihydro-2H-iImidazole-​2-​thione, BIA 5-453

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C14H15F2N3OS
CAS Number:
Molecular Weight:
311.35
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
eCl@ss:
32150313
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

Orally active, potent, reversible and peripherally selective dopamine-β-hydroxylase (DBH) inhibitor.
Etamicastat (BIA 5-453) is an orally active, potent, reversible and peripherally selective dopamine-β-hydroxylase (DBH) inhibitor that reduces both systolic (SBP) and diastolic (DBP) blood pressure.


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library