Skip to Content
MilliporeSigma

SML3860

Conolidine

≥98% (HPLC)

Synonym(s):

(+)-Conolidine, (+)-Ervaticine, (2R,4E,5S)-4-Ethylidene-1,4,5,7-tetrahydro-2,5-ethano-2H-azocino[4,3-b]indol-6(3H)-one, Ervaticine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C17H18N2O
CAS Number:
Molecular Weight:
266.34
UNSPSC Code:
51111800
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: >2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

O=C(C1=C(C[N@](CC2)C/3)C4=C(C=CC=C4)N1)[C@@H]2C3=C(C)\[H]

InChI

1S/C17H18N2O/c1-2-11-9-19-8-7-12(11)17(20)16-14(10-19)13-5-3-4-6-15(13)18-16/h2-6,12,18H,7-10H2,1H3/b11-2-/t12-/m0/s1

InChI key

DBGBUYFOJXOYNY-RENATIMJSA-N

Biochem/physiol Actions

Natural analgesic that induces pain relieve though activation of atypical scavenger receptor for chemokines and opioid peptides ACKR3 (CXCR7).



Conolidine is a natural analgesic isolated from tropical flowering shrub Tabernaemontana divaricate, which exhibits potent analgesia in in vivo models of tonic and persistent pain. Conolidine induces pain relieve though activation of atypical scavenger receptor for chemokines and opioid peptides ACKR3 (CXCR7). It appears that conolidine inhibits scavenging functions of ACKR3 therefore increasing levels of endogenous opioid peptides.


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

WGK

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library