Skip to Content
MilliporeSigma

Skip To

SML3868

PG3d

≥97% (1H-NMR)

Synonym(s):

4-Hydroxy-2-(2-phenylethynyl)-5-(phenylmethyl)-6H-1,3-oxazin-6-one, 5-Benzyl-4-hydroxy-2-(phenylethynyl)-6H-1,3-oxazin-6-one

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C19H13NO3
CAS Number:
Molecular Weight:
303.31
UNSPSC Code:
12352200
NACRES:
NA.21
MDL number:
Assay:
≥97% (1H-NMR)
Form:
powder
Storage condition:
desiccated
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥97% (1H-NMR)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

N1=C(OC(=O)C(C1=O)Cc3ccccc3)C#Cc2ccccc2

InChI

1S/C19H13NO3/c21-18-16(13-15-9-5-2-6-10-15)19(22)23-17(20-18)12-11-14-7-3-1-4-8-14/h1-10,16H,13H2

InChI key

ZBZZHVGCNWRIGR-UHFFFAOYSA-N

Biochem/physiol Actions

Cell penetrant, potent and selective irreversible allosteric inhibitor of caspase-6 activation that binds outside the active site of caspase-6

Disclaimer

Hygroscopic


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters